missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',4'-Dimethylacetophenone, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.002 |
| Boiling Point | 242°C to 243°C |
| Flash Point | 102°C (215°F) |
| Refractive Index | 1.538 |
| Quantity | 5 g |
| Beilstein | 907053 |
| Synonym | 3',4'-dimethylacetophenone, 1-3,4-dimethylphenyl ethanone, 3,4-dimethylacetophenone, ethanone, 1-3,4-dimethylphenyl, 1-3,4-dimethylphenyl ethan-1-one, unii-pc1l145t5v, 1-3,4-dimethyl-phenyl-ethanone, 1-acetyl-3,4-dimethylbenzene, 3.4-dimethylacetophenon, acmc-1cmy2 |
| SMILES | CC1=C(C=C(C=C1)C(=O)C)C |
| IUPAC Name | 1-(3,4-dimethylphenyl)ethanone |
| PubChem CID | 77193 |
| Show More |
Safety and Handling
Product Identifier
- 3',4'-Dimethylacetophenone
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: 1-(3,4-dimethylphenyl)ethanone
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?