missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethylacetophenone, 98%

Product Code. 11454833
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454833 25 g 25g
11444833 5 g 5g
11464833 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454833 Supplier Thermo Scientific Alfa Aesar Supplier No. A12187.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3637-01-2 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3637-01-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00008743
InChI Key WPRAXAOJIODQJR-UHFFFAOYSA-N

Specifications

Density 1.002
Boiling Point 242°C to 243°C
Flash Point 102°C (215°F)
Refractive Index 1.538
Quantity 25 g
Beilstein 907053
Synonym 3',4'-dimethylacetophenone, 1-3,4-dimethylphenyl ethanone, 3,4-dimethylacetophenone, ethanone, 1-3,4-dimethylphenyl, 1-3,4-dimethylphenyl ethan-1-one, unii-pc1l145t5v, 1-3,4-dimethyl-phenyl-ethanone, 1-acetyl-3,4-dimethylbenzene, 3.4-dimethylacetophenon, acmc-1cmy2
SMILES CC1=C(C=C(C=C1)C(=O)C)C
IUPAC Name 1-(3,4-dimethylphenyl)ethanone
PubChem CID 77193
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 3',4'-Dimethylacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.