missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-2-hexanol, erythro + threo, 98%

Product Code. 11439716
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11439716

Brand: Thermo Scientific Alfa Aesar B20243.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19550-05-1 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19550-05-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00021887
InChI Key LBJWJHMCZHKLQU-UHFFFAOYSA-N
Synonym 3,4-dimethyl-2-hexanol, 2-hexanol, 3,4-dimethyl, 3,4-dimethyl-2-hexanol, erythro + threo, 3,4-dimethyl-2-hexanol mixture of isomers, 2-hexanol,4-dimethyl, acmc-1ah66, 3,4-dimethyl-2-hexanol mixture of isomers a, 3,4-dimethyl-2-hexanol, threo + erythro
PubChem CID 140547
IUPAC Name 3,4-dimethylhexan-2-ol
SMILES CCC(C)C(C)C(C)O

Specifications

Quantity 25 g
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 3,4-Dimethyl-2-hexanol, erythro + threo

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.