missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-2(5H)-furanone, TRC

Product Code. 30385608
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385608 1 g
30381564 250 mg
30376187 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385608 Supplier Toronto Research Chemicals Supplier No. TRCD4777651G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1575-46-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
Synonym NSC 362902, 2,3-Dimethyl-2-butenoic Acid gamma-Lactone, 2,3-Dimethyl-4-hydroxy-2-butenoic Acid Lactone, 2,3-Dimethyl-4-hydroxy-2-butenoic Acid gamma-Lactone, 3,4-Dimethyl-2(5H)-furanone
IUPAC Name 3,4-dimethyl-2H-furan-5-one
SMILES CC1=C(C)C(=O)OC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H8O2/c1-4-3-8-6(7)5(4)2/h3H2,1-2H3
Formula Weight 112.0524
Chemical Name or Material 3,4-Dimethyl-2(5H)-furanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.