missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-1H-pyrazole, 97%

Product Code. p-7052575
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447737 5 g 5g
15457737 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447737 Supplier Thermo Scientific Alfa Aesar Supplier No. H37099.06
Add to basket
This item is not returnable. View return policy

CAS: 2820-37-3 | C5H8N2 | 96.133 g/mol

3,4-Dimethyl-1H-pyrazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethyl-1H-pyrazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2820-37-3
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.133
MDL Number MFCD00462154
InChI Key VQTVFIMEENGCJA-UHFFFAOYSA-N
Synonym 3,4-dimethyl-1h-pyrazole, 3 5 ,4-dimethylpyrazole, 3,4-dimethylpyrazole, 3,4-dimethyl pyrazole, 1h-pyrazole, 3,4-dimethyl, unii-7lf1k5quj0, 7lf1k5quj0, 1h-pyrazole,4,5-dimethyl, 1h-pyrazole, 4,5-dimethyl, pyrazole, 3,4-dimethyl
PubChem CID 137735
IUPAC Name 4,5-dimethyl-1H-pyrazole
SMILES CC1=C(NN=C1)C

Specifications

Melting Point 53°C to 55°C
Color Cream
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 96.13
Percent Purity 97%
Physical Form Fused Solid
Chemical Name or Material 3,4-Dimethyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.