missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-1,2-cyclopentanedione, 97%, Thermo Scientific™

Product Code. 10083370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
2.5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10083370 2.5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083370 Supplier Thermo Scientific Acros Supplier No. 265050025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 13494-06-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 64°C to 69°C
Color Yellow to Beige
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H10O2
MDL Number MFCD00143063
Quantity 2.5 g
Synonym 3,4-dimethyl-1,2-cyclopentanedione, 3,4-dimethyl-1,2-cyclopentadione, 1,2-cyclopentanedione, 3,4-dimethyl, fema no. 3268, 1,2-cyclopentanedione,3,4-dimethyl, caramel pentadione, 3,4-dimethyl 1,2-cyclopentandione, acmc-209bxe, 3,4-dimethyl-1,2-cyclopentadione, fg
Solubility Information Solubility in water: slightly soluble
InChI Key WGAVDEVFJDQIMZ-UHFFFAOYSA-N
SMILES CC1CC(=O)C(=O)C1C
IUPAC Name 3,4-dimethylcyclopentane-1,2-dione
Molecular Weight (g/mol) 126.15
PubChem CID 61633
Formula Weight 126.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dimethyl-1, 2-cyclopentanedione, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.