missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxythiophene, 98%

Product Code. 15473188
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473188 1 g 1g
15483188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473188 Supplier Thermo Scientific Alfa Aesar Supplier No. H56674.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 51792-34-8 | C6H8O2S | 144.188 g/mol

3,4-Dimethoxythiophene is used as an electronic materials intermediate. It is used as a starting material in the synthesis of porphyrin dyad which is used to study photoinduced energy transfer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxythiophene is used as an electronic materials intermediate. It is used as a starting material in the synthesis of porphyrin dyad which is used to study photoinduced energy transfer.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51792-34-8
Molecular Formula C6H8O2S
Molecular Weight (g/mol) 144.188
MDL Number MFCD01096546
InChI Key ZUDCKLVMBAXBIF-UHFFFAOYSA-N
Synonym 3,4-dimethoxy thiophene, thiophene, 3,4-dimethoxy, pubchem15752, 3,4-dimethoxy-thiophene, acmc-209kw1, ksc491i4n, 3,4-dimethoxythiophene
PubChem CID 3613501
IUPAC Name 3,4-dimethoxythiophene
SMILES COC1=CSC=C1OC

Specifications

Density 1.209
Boiling Point 100°C to 102°C (10 mmHg to 11 mmHg)
Flash Point 106°C (223°F)
Refractive Index 1.5409
Quantity 1 g
Solubility Information Miscible with organic solvents. Slightly miscible with water.
Formula Weight 144.19
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxythiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.