missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylglyoxal hydrate, 98%, dry wt basis

Product Code. 11389146
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389146 1 g 1g
11399146 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389146 Supplier Thermo Scientific Alfa Aesar Supplier No. L20279.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 163428-90-8 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163428-90-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD03411519
InChI Key QQSUAMUBZNQYGE-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylglyoxal hydrate, 2-3,4-dimethoxyphenyl-2-oxoacetaldehyde hydrate, pubchem12147, acmc-1c6zz, 3,4-dimethoxyphenylglyoxalhydrate, 2-3,4-dimethoxyphenyl-2-oxo-acetaldehyde, 2-3,4-dimethoxyphenyl-2-oxoethanal, hydrate, ethanone,1-3,4-dimethoxyphenyl-2,2-dihydroxy, 3,4-dimethoxyphenyl oxo acetaldehyde-water 1/1, 3,4-dimethoxyphenyl oxo acetaldehyde hydrate 1:1
PubChem CID 44118538
IUPAC Name 2-(3,4-dimethoxyphenyl)-2-oxoacetaldehyde;hydrate
SMILES COC1=CC=C(C=C1OC)C(=O)C=O

Specifications

Melting Point 118°C to 120°C
Quantity 1 g
Beilstein 3092619
Formula Weight 194.19 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 3,4-Dimethoxyphenylglyoxal hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.