missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetonitrile, Mikromol™

Product Code. 30049724
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049724 100 mg 100mg
30051489 25 mg 25mg
30458474 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30049724 Supplier Mikromol Supplier No. MM0022.18

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,4-Dimethoxyphenylacetonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dimethoxyphenylacetonitrile
CAS 93-17-4
Analyte or Component Names 3,4-Dimethoxyphenylacetonitrile
Shipping Condition Room Temperature
Molecular Formula C10 H11 N O2
Certifications/Compliance ISO 17025
Synonym 3,4-Dimethoxyphenylacetonitrile
InChI Formula InChI=1S/C10H11NO2/c1-12-9-4-3-8(5-6-11)7-10(9)13-2/h3-4,7H,5H2,1-2H3
SMILES COc1ccc(CC#N)cc1OC
IUPAC Name 2-(3,4-dimethoxyphenyl)acetonitrile
Molecular Weight (g/mol) 177.2
Quantity 100 mg
Formula Weight 177.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.