missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetone, 97%

Product Code. 11327465
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327465 50 g 50g
11317465 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327465 Supplier Thermo Scientific Alfa Aesar Supplier No. L05477.18
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 776-99-8 | C11H14O3 | 194.23 g/mol

3,4-Dimethoxyphenylacetone was used as starting material in the synthesis of α-methyl-dopa, an important nonproteogenic α-amino acid for pharmaceutical applications. 3,4-Dimethoxyphenylacetone undergoes asymmetric amination catalyzed by Brevibacterium linens IFO 12141 strain to yield corresponding optically active amine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxyphenylacetone was used as starting material in the synthesis of α-methyl-dopa, an important nonproteogenic α-amino acid for pharmaceutical applications. 3,4-Dimethoxyphenylacetone undergoes asymmetric amination catalyzed by Brevibacterium linens IFO 12141 strain to yield corresponding optically active amine

Solubility
Insoluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776-99-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008772
InChI Key UMYZWICEDUEWIM-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
PubChem CID 69896
IUPAC Name 1-(3,4-dimethoxyphenyl)propan-2-one
SMILES COC1=CC=C(CC(C)=O)C=C1OC

Specifications

Density 1.115
Boiling Point 154°C to 158°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5358
Quantity 50 g
Beilstein 1107410
Solubility Information Insoluble in water.
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxyphenylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.