missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetone, 97%, Thermo Scientific™

Product Code. 15535046 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15535046 5 g 5g
15545046 25 g 25g
15555046 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15535046 Supplier Thermo Scientific Acros Supplier No. 161490050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776-99-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008772
InChI Key UMYZWICEDUEWIM-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
PubChem CID 69896
IUPAC Name 1-(3,4-dimethoxyphenyl)propan-2-one
SMILES COC1=CC=C(CC(C)=O)C=C1OC

Specifications

Density 1.1150g/mL
Boiling Point 154.0°C to 158.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula (CH3O)2C6H3CH2COCH3
Refractive Index 1.5338 to 1.5358
Quantity 5 g
Specific Gravity 1.115
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxyphenylacetone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.