missing translation for 'onlineSavingsMsg'
Learn More

(3,4-Dimethoxyphenyl)acetonitrile, 99+%

Product Code. 10571374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571374 500 g Glass bottle 500g
10021780 25 g Glass bottle 25g
10206160 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10571374 Supplier Thermo Scientific Acros Supplier No. 155265000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-17-4 | C10H11NO2 | 177.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-17-4
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00001911
InChI Key ASLSUMISAQDOOB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile
PubChem CID 66727
IUPAC Name 2-(3,4-dimethoxyphenyl)acetonitrile
SMILES COC1=C(C=C(C=C1)CC#N)OC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 62°C to 65°C
Color White
Boiling Point 171°C to 178°C (10.0 mmHg)
Flash Point 250°C
Loss on Drying 0.5% max. (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2CN
Quantity 500 g
Beilstein 10, I, 198
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,chloroform and,ethanol
Formula Weight 177.2
Percent Purity 99+%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material (3, 4-Dimethoxyphenyl)acetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • (3,4-Dimethoxyphenyl)acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.