missing translation for 'onlineSavingsMsg'
Learn More

(3,4-Dimethoxyphenyl)(2-(2-hydrazonopentan-3-yl)-4,5-dimethoxyphenyl)methanone, TRC

Product Code. 30384616
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384616 10 mg -
30377570 50 mg 50mg
30379298 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384616 Supplier Toronto Research Chemicals Supplier No. TRCD47062610MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37952-09-3
Molecular Formula C22H28N2O5
Molecular Weight (g/mol) 400.47
Synonym Tofisopam impurity
SMILES CCC(\C(=N\N)\C)c1cc(OC)c(OC)cc1C(=O)c2ccc(OC)c(OC)c2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H28N2O5/c1-7-15(13(2)24-23)16-11-20(28-5)21(29-6)12-17(16)22(25)14-8-9-18(26-3)19(10-14)27-4/h8-12,15H,7,23H2,1-6H3/b24-13-
Formula Weight 400.1998
Chemical Name or Material (3,4-Dimethoxyphenyl)(2-(2-hydrazonopentan-3-yl)-4,5-dimethoxyphenyl)methanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.