missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenethyl isothiocyanate, 95%, Maybridge™

Brand:  Maybridge TL00669DA

Code : 31

Additional Details : CAS Number : 21714-25-0 Weight : 0.05100kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 10007982

  • £13.06 / 1g

Estimated Shipment
23-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

21714-25-0
223.29
3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
C11H13NO2S
GHYFLDWHIVKQLS-UHFFFAOYSA-N
140856
COC1=C(C=C(C=C1)CCN=C=S)OC

Specifications

3, 4-Dimethoxyphenethyl isothiocyanate
21714-25-0
C11H13NO2S
GHYFLDWHIVKQLS-UHFFFAOYSA-N
4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
140856
1g
95%
3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
COC1=C(C=C(C=C1)CCN=C=S)OC
223.29
223.29
Videos
SDS Documents