missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxycinnamic acid, MedChemExpress

Product Code. 16469166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469166 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469166 Supplier Medchem Express Supplier No. HYN1778
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3,4-Dimethoxycinnamic acid (O-Methylferulic acid) is a monomer extracted and purified from Securidaca inappendiculata Hassk. 3,4-Dimethoxycinnamic acid exerts anti-apoptotic effects on L-02 cells via the ROS-mediated signaling pathway. Anti-apoptotic effects.

3,4-Dimethoxycinnamic acid (O-Methylferulic acid) is a monomer extracted and purified from Securidaca inappendiculata Hassk. 3,4-Dimethoxycinnamic acid exerts anti-apoptotic effects on L-02 cells via the ROS-mediated signaling pathway. Anti-apoptotic effects.

Specifications

Chemical Name or Material 3,4-Dimethoxycinnamic acid
CAS 2316-26-9
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C11H12O4
Quantity 100 mg
Synonym O-Methylferulic acid
Solubility Information DMSO : 50 mg/mL (240.14 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C1=CC=C(OC)C(OC)=C1
Molecular Weight (g/mol) 208.21
Formula Weight 208.21
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.