missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzylamine, 97%

Product Code. 10055763 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055763 100 g Glass bottle 100g
10672382 5 g Glass bottle 5g
10439281 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10055763 Supplier Thermo Scientific Acros Supplier No. 140831000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5763-61-1 | C9H13NO2 | 167.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5763-61-1
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00008116
InChI Key DIVNUTGTTIRPQA-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
PubChem CID 79832
IUPAC Name (3,4-dimethoxyphenyl)methanamine
SMILES COC1=CC=C(CN)C=C1OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1000g/mL
Boiling Point 281°C to 284°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2NH2
Refractive Index 1.5530 to 1.557
Quantity 100 g
Beilstein 13, 796
Specific Gravity 1.1
Formula Weight 167.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.