missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxybenzyl Alcohol, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | (3,4-Dimethoxyphenyl)-methanol |
| CAS | 93-03-8 |
| Molecular Formula | C9 H12 O3 |
| Quantity | 100 g |
| Synonym | (3,4-Dimethoxyphenyl)-methanol, Verapamil Hydrochloride Imp. E (EP) |
| InChI Formula | InChI=1S/C9H12O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-5,10H,6H2,1-2H3 |
| SMILES | COc1ccc(CO)cc1OC |
| IUPAC Name | (3,4-dimethoxyphenyl)methanol |
| Molecular Weight (g/mol) | 168.19 |
| Formula Weight | 168.0786 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction