missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzoyl chloride, 98%

Product Code. 10668211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10668211 5 g Glass bottle 5g
10239553 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10668211 Supplier Thermo Scientific Acros Supplier No. 305600050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3535-37-3 | C9H9ClO3 | 200.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3535-37-3
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.62
MDL Number MFCD00000674
InChI Key VIOBGCWEHLRBEP-UHFFFAOYSA-N
Synonym veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t
PubChem CID 77070
IUPAC Name 3,4-dimethoxybenzoyl chloride
SMILES COC1=C(C=C(C=C1)C(=O)Cl)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 68.0°C to 72.0°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3COCl
Quantity 5 g
Formula Weight 200.62
Percent Purity 98%
Physical Form Crystals, Flakes or Chunks
Chemical Name or Material 3, 4-Dimethoxybenzoyl chloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.