missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzonitrile, 98+%

Product Code. 11484174
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484174 25 g 25g
11494174 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484174 Supplier Thermo Scientific Alfa Aesar Supplier No. A12521.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2024-83-1 | C9H9NO2 | 163.18 g/mol

3,4-Dimethoxybenzonitrile is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzonitrile is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2024-83-1
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00001802
InChI Key OSEQIDSFSBWXRE-UHFFFAOYSA-N
Synonym veratronitrile, 4-cyanoveratrole, 3,4-dimethoxy-benzonitrile, 4-cyano-1,2-dimethoxybenzene, 1-cyano-3,4-dimethoxylbenzene, 3,4-dimethoxybenzoic acid nitrile, 3,4-dimethoxyphenyl cyanide, benzonitrile,4-dimethoxy, 3,4-dimethoxybenzenecarbonitrile, 3-cyanoveratrole
PubChem CID 74842
IUPAC Name 3,4-dimethoxybenzonitrile
SMILES COC1=CC=C(C=C1OC)C#N

Specifications

Melting Point 65°C to 69°C
Quantity 25 g
UN Number UN3439
Beilstein 1451651
Formula Weight 163.18
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethoxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.