missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzoic Acid (Veratric Acid), Mikromol™

Product Code. 17900646
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17900646 25 mg 25mg
17900496 100 mg 100mg
30457408 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17900646 Supplier Mikromol Supplier No. MM0319.010025
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,4-Dimethoxybenzoic Acid (Veratric Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Veratric Acid
CAS 93-07-2
Analyte or Component Names Veratric Acid
Shipping Condition Room Temperature
Molecular Formula C9 H10 O4
Certifications/Compliance ISO 17025
Synonym 3,4-Dimethoxybenzoic Acid, Mebeverine Hydrochloride Imp. D (Pharmeuropa), Veratric Acid, Mebeverine Hydrochloride Impurity D, Mebeverine Impurity D
InChI Formula InChI=1S/C9H10O4/c1-12-7-4-3-6(9(10)11)5-8(7)13-2/h3-5H,1-2H3,(H,10,11)
SMILES COc1ccc(cc1OC)C(=O)O
IUPAC Name 3,4-dimethoxybenzoic acid
Molecular Weight (g/mol) 182.17
Quantity 25 mg
Formula Weight 182.0579 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.