missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%

Product Code. 11459453
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11459453

Brand: Thermo Scientific Alfa Aesar A12337.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 328-70-1 | C8H3BrF6 | 293.006 g/mol

1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-70-1
Molecular Formula C8H3BrF6
Molecular Weight (g/mol) 293.006
MDL Number MFCD00000381
InChI Key CSVCVIHEBDJTCJ-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl bromobenzene, 1-bromo-3,5-bis trifluoromethyl benzene, 1,3-bis trifluoromethyl-5-bromobenzene, mbt-br, 3,5-di trifluoromethyl bromobenzene, 3,5-bis trifluoromethyl-1-bromobenzene, 3,5-bis-trifluoromethylbromobenzene, benzene, 1-bromo-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl bromide, 1-bromo-3,5-bis-trifluoromethyl-benzene
PubChem CID 67602
IUPAC Name 1-bromo-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)Br)C(F)(F)F

Specifications

Melting Point -16°C
Density 1.698
Boiling Point 152°C to 155°C
Flash Point >110°C (230°F)
Refractive Index 1.427
Quantity 10 g
Beilstein 2123669
Solubility Information Immiscible with water.
Formula Weight 293.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-3,5-bis (trifluoromethyl)benzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.