Learn More
3,4-Dimethoxybenzoic acid, 99+%
Description
3,4-Dimethoxybenzoic acid plays an important role in producing antibiotics and various dyes. They are used as intermediates for pharmaceuticals (especially for antipyretic analgesic, antirheumatism) and other organic synthesis. It is used as matrix for ionization of peptides, proteins and carbohydrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,4-Dimethoxybenzoic acid plays an important role in producing antibiotics and various dyes. They are used as intermediates for pharmaceuticals (especially for antipyretic analgesic, antirheumatism) and other organic synthesis. It is used as matrix for ionization of peptides, proteins and carbohydrates.
Solubility
Soluble in water (0.5 mg/ml at 15°C), chloroform, methanol, and 95% ethanol (50 mg/ml).
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Specifications
| Melting Point | 179°C to 183°C |
| Odor | Odorless |
| Quantity | 1000 g |
| Beilstein | 518285 |
| Merck Index | 14,9953 |
| Solubility Information | Soluble in water (0.5mg/ml at 15°C),chloroform,methanol,and 95% ethanol (50mg/ml). |
| Formula Weight | 182.18 |
| Percent Purity | ≥99% |
| Chemical Name or Material | 3,4-Dimethoxybenzoic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.