missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzenesulfonyl chloride, 97%

Product Code. 10652223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10652223 5 g Glass bottle 5g
10363962 1 g Glass bottle 1g
10095863 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10652223 Supplier Thermo Scientific Acros Supplier No. 354390050
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 23095-31-0 | C8H9ClO4S | 236.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23095-31-0
Molecular Formula C8H9ClO4S
Molecular Weight (g/mol) 236.68
MDL Number MFCD00051769
InChI Key RSJSYCZYQNJQPY-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzene-1-sulfonyl chloride, 3,4-dimethoxybenzenesulfonylchloride, 3,4-dimethoxyphenylsulfonyl chloride, 3,4-dimethoxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethoxy, 3,4-dimethoxyphenyl chlorosulfone, pubchem5127, acmc-1co88, asischem d48920, zerenex zx009571
PubChem CID 2734183
IUPAC Name 3,4-dimethoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 65.0°C to 70.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3SO2Cl
Quantity 5 g
Formula Weight 236.68
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dimethoxybenzenesulfonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.