missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water (25 g/L).
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water (25 g/L).
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | ∼240°C |
Quantity | 5 g |
Beilstein | 5334877 |
Solubility Information | Soluble in water (25g/L). |
Formula Weight | 181.98 |
Percent Purity | 98% |
Chemical Name or Material | 3,4-Dimethoxybenzeneboronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'