missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzaldehyde, TRC

Product Code. 30012180
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012180 5 g 5g
30010600 10 g 10g
17902745 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012180 Supplier Toronto Research Chemicals Supplier No. TRCD4606635G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-14-9
Molecular Formula C9 H10 O3
Molecular Weight (g/mol) 166.17
Synonym 3,4-Dimethoxybenzaldehyde, Verapamil Hydrochloride Imp. G (EP), Benzaldehyde, 3,4-dimethoxy-, Veratraldehyde (7CI,8CI), 3,4-Dimethoxybenzenecarbonal, 3,4-Dimethoxyphenylaldehyde, 4-O-Methylvanillin, Methylvanillin, NSC 24521, NSC 8500, Protocatechualdehyde dimethyl ether, Protocatechuic aldehyde dimethyl ether, Vanillin methyl ether, Veratral, Veratric aldehyde, Veratrum aldehyde, Veratryl aldehyde
IUPAC Name 3,4-dimethoxybenzaldehyde
SMILES COc1ccc(C=O)cc1OC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H10O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-6H,1-2H3
Formula Weight 166.063
Chemical Name or Material 3,4-Dimethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.