missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzaldehyde, 99+%

Product Code. 10275540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275540 100 g Plastic bottle 100g
10245590 5 g Glass Bottle 5g
10144320 500 g Plastic bottle 500g
10628064 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10275540 Supplier Thermo Scientific Acros Supplier No. 162611000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-14-9 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-14-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00003363
InChI Key WJUFSDZVCOTFON-UHFFFAOYSA-N
Synonym veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin
PubChem CID 8419
ChEBI CHEBI:17098
IUPAC Name 3,4-dimethoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1OC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 42°C to 45°C
Color White to Yellow
Boiling Point 281°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3O)2C6H3CHO
Quantity 100 g
Beilstein 08, 255
Merck Index 15, 10145
Solubility Information Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether
Formula Weight 166.18
Percent Purity 99+%
Physical Form Lumps or Fused Solid
Chemical Name or Material 3, 4-Dimethoxybenzaldehyde, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.