missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxyaniline, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.
Solubility
Slightly soluble in water
Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.
Solubility
Slightly soluble in water
Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | 86°C to 89°C |
Boiling Point | 174°C to 176°C (22 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Characteristic |
Quantity | 50 g |
UN Number | UN2811 |
Beilstein | 743399 |
Solubility Information | Slightly soluble in water |
Formula Weight | 153.18 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'