missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyaniline, 98+%

Product Code. 11440034
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440034 10 g 10g
11450034 50 g 50g
11460034 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440034 Supplier Thermo Scientific Alfa Aesar Supplier No. A13548.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6315-89-5 | C8H11NO2 | 153.181 g/mol

3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.

Solubility
Slightly soluble in water

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6315-89-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00008394
InChI Key LGDHZCLREKIGKJ-UHFFFAOYSA-N
Synonym 4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
PubChem CID 22770
IUPAC Name 3,4-dimethoxyaniline
SMILES COC1=C(C=C(C=C1)N)OC

Specifications

Melting Point 86°C to 89°C
Boiling Point 174°C to 176°C (22 mmHg)
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 10 g
UN Number UN2811
Beilstein 743399
Solubility Information Slightly soluble in water
Formula Weight 153.18
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethoxyaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.