missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyacetophenone, 98+%

Product Code. 11337815
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337815 50 g 50g
11347815 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337815 Supplier Thermo Scientific Alfa Aesar Supplier No. L02737.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1131-62-0 | C10H12O3 | 180.203 g/mol

3',4'-Dimethoxyacetophenone is used as Catalytic agent; Petrochemical additive. It is also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′-Dimethoxyacetophenone is used as Catalytic agent; Petrochemical additive. It is also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Protect from moisture, heat. Incompatible with Strong oxidizing agents. Protect from direct light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-62-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00008737
InChI Key IQZLUWLMQNGTIW-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
PubChem CID 14328
ChEBI CHEBI:86576
IUPAC Name 1-(3,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)OC

Specifications

Melting Point 47°C to 51°C
Boiling Point 286°C to 288°C
Flash Point >110°C (230°F)
Odor Odorless
Quantity 50 g
Beilstein 781213
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 3',4'-Dimethoxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.