missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyacetophenone, 98%

Product Code. 10275930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275930 100 g Glass bottle 100g
10612413 5 g Glass bottle 5g
10406432 25 g Glass bottle 25g
10076093 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10275930 Supplier Thermo Scientific Acros Supplier No. 168851000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1131-62-0 | C10H12O3 | 180.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-62-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00008737
InChI Key IQZLUWLMQNGTIW-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
PubChem CID 14328
ChEBI CHEBI:86576
IUPAC Name 1-(3,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48.0°C to 52.0°C
Color Yellow to Beige
Boiling Point 286.0°C to 288.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3COCH3
Quantity 100 g
Beilstein 08, II, 298
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol
Formula Weight 180.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3', 4'-Dimethoxyacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 3',4'-Dimethoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.