missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy Styrene (~1% Hydroquinone as stabilizer), TRC

Product Code. 30388330
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388330 25 g -
30384804 10 g 10g
30377785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388330 Supplier Toronto Research Chemicals Supplier No. TRCD46051525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6380-23-0
Molecular Formula C10 H12 O2
Molecular Weight (g/mol) 164.2
Synonym Benzene, 4-ethenyl-1,2-dimethoxy-, Styrene, 3,4-dimethoxy- (7CI,8CI), Veratrole, 4-vinyl- (6CI), 4-Ethenyl-1,2-dimethoxybenzene, 1,2-Dimethoxy-4-vinylbenzene, 3,4-Dimethoxystyrene, 3,4-Dimethoxystyrol, 4-Vinyl-1,2-dimethoxybenzene, 4-Vinylveratrole
IUPAC Name 4-ethenyl-1,2-dimethoxybenzene
SMILES COc1ccc(C=C)cc1OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H12O2/c1-4-8-5-6-9(11-2)10(7-8)12-3/h4-7H,1H2,2-3H3
Formula Weight 164.0837
Chemical Name or Material 3,4-Dimethoxy Styrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.