missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy-Alpha-(1-methylethyl-d7)benzeneacetonitrile, TRC

Product Code. 30379816
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379816 10 mg
30388039 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30379816 Supplier Toronto Research Chemicals Supplier No. TRCD47478210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C13H10D7NO2
Molecular Weight (g/mol) 226.33
Synonym (±)-2-(3,4-Dimethoxyphenyl)-3-methylbutane-d7-nitrile, (±)-3,4-Dimethoxy-α-isopropyl-d7-benzeneacetonitrile, 2-(3,4-Dimethoxyphenyl)-2-isopropyl-d7-acetonitrile, 2-(3,4-Dimethoxyphenyl)-3-methylbutane-d7-nitrile, 2-(3,4-Dimethoxyphenyl)-3-methylbutyro-d7-nitrile, 3,4-Dimethoxy-α-(1-methylethyl-d7)benzeneacetonitrile, Isopropyl-d7-(3,4-dimethoxyphenyl)acetonitrile, α-Isopropyl-d7-homoveratronitrile, α-Isopropyl-d7-veratryl cyanide
IUPAC Name 2-(3,4-dimethoxyphenyl)-3-(methyl-d3)butanenitrile-3,4,4,4-d4
SMILES [2H]C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C(C#N)C1=CC(OC)=C(OC)C=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H17NO2/c1-9(2)11(8-14)10-5-6-12(15-3)13(7-10)16-4/h5-7,9,11H,1-4H3/i1D3,2D3,9D
Formula Weight 226.17
Chemical Name or Material 3,4-Dimethoxy-alpha-(1-methylethyl-d7)benzeneacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.