missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy[7-13C]-benzaldehyde, TRC

Product Code. 30386725
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386725 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386725 Supplier Toronto Research Chemicals Supplier No. TRCD46066250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dimethoxy[7-13C]-benzaldehyde
CAS 1173022-44-0
Molecular Formula 13C C8 H10 O3
Quantity 50 mg
Synonym 3,4-Dimethoxybenzaldehyde-13C, 3,4-Dimethoxybenzaldehyde-7-13C, 3,4-Dimethoxy[7-13C]-benzaldehyde, 3,4-Dimethoxybenzaldehyde-13C (carbonyl-13C), Veratraldehyde-13C, 4-O-Methylvanillin-13C, Vanillin-13C methyl ether
InChI Formula InChI=1S/C9H10O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-6H,1-2H3/i6+1
SMILES COc1ccc([13CH]=O)cc1OC
IUPAC Name 3,4-dimethoxybenzaldehyde
Molecular Weight (g/mol) 167.17
Formula Weight 167.0663
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.