missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dihydroxypropiophenone, 98%

Product Code. 11425503
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425503 5 g 5g
11415503 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425503 Supplier Thermo Scientific Alfa Aesar Supplier No. A13444.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7451-98-1 | C9H10O3 | 166.176 g/mol

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7451-98-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016440
InChI Key HNWIHBDMOYWCGX-UHFFFAOYSA-N
Synonym 3',4'-dihydroxypropiophenone, 3,4-dihydroxypropiophenone, propiophenone, 3',4'-dihydroxy, 1-3,4-dihydroxyphenyl propan-1-one, 1-propanone, 1-3,4-dihydroxyphenyl, 1-3,4-dihydroxyphenyl-1-propanone, 1-propanone,1-3,4-dihydroxyphenyl, 4-propionylbrenzcatechin, propiophenone,4'-dihydroxy, 3', 4'-dihydroxypropiophenone
PubChem CID 96267
IUPAC Name 1-(3,4-dihydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC(=C(C=C1)O)O

Specifications

Melting Point 145°C to 146°C
Quantity 5 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3',4'-Dihydroxypropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.