missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dihydroxyphenylacetone, TRC

Product Code. 30008703
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008703 25 mg 25mg
30007289 100 mg 100mg
30003518 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008703 Supplier Toronto Research Chemicals Supplier No. TRCD45430025MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2503-44-8
Molecular Formula C9 H10 O3
Molecular Weight (g/mol) 166.17
Synonym 1-(3,4-Dihydroxyphenyl)-2-propanone, 2-Propanone, (3,4-dihydroxyphenyl)- (6CI,7CI), 1-(3,4-Dihydroxyphenyl)-2-propanone, 1-(3',4'-Dihydroxyphenyl)-2-propanone, 3,4-Dihydroxyphenylacetone, Carbidopa Imp. G (EP)
IUPAC Name 1-(3,4-dihydroxyphenyl)propan-2-one
SMILES CC(=O)Cc1ccc(O)c(O)c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H10O3/c1-6(10)4-7-2-3-8(11)9(12)5-7/h2-3,5,11-12H,4H2,1H3
Formula Weight 166.063
Percent Purity >95%
Chemical Name or Material 3,4-Dihydroxyphenylacetone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.