missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxyphenylacetic Acid, TRC

Product Code. 30012348
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012348 25 g 25g
30003549 5 g 5g
30008310 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012348 Supplier Toronto Research Chemicals Supplier No. TRCD45425025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-32-9
Molecular Formula C8 H8 O4
Molecular Weight (g/mol) 168.15
Synonym Benzeneacetic acid, 3,4-dihydroxy-, Acetic acid, (3,4-dihydroxyphenyl)- (8CI), 3,4-Dihydroxybenzeneacetic acid, (3,4-Dihydroxyphenyl)acetic acid, 3',4'-Dihydroxyphenylacetic acid, Ba 2773, DOPAC, Dihydroxyphenylacetic acid, Dopacetic acid, Homoprotocatechuic acid, NSC 73191
IUPAC Name 2-(3,4-dihydroxyphenyl)acetic acid
SMILES OC(=O)Cc1ccc(O)c(O)c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H8O4/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3,9-10H,4H2,(H,11,12)
Formula Weight 168.0423
Chemical Name or Material 3,4-Dihydroxyphenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.