missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzylamine hydrobromide, 98%

Product Code. 10338650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10338650 5 g Glass bottle 5g
10561861 250 mg Glass bottle 250mg
10206380 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10338650 Supplier Thermo Scientific Acros Supplier No. 228570050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16290-26-9 | C7H10BrNO2 | 220.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16290-26-9
Molecular Formula C7H10BrNO2
Molecular Weight (g/mol) 220.07
MDL Number MFCD00012859
InChI Key BVFZTXFCZAXSHN-UHFFFAOYSA-N
Synonym 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, dhba hydrobromide, acmc-1afa4, 1, 4-aminomethyl-, hydrobromide, 4-aminomethyl catechol hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide
PubChem CID 13343562
IUPAC Name 4-(aminomethyl)benzene-1,2-diol;hydrobromide
SMILES [H+].[Br-].NCC1=CC=C(O)C(O)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >180.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (HO)2C6H3CH2NH2·HBr
Quantity 5 g
Beilstein 13, 796
Formula Weight 220.07
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dihydroxybenzylamine hydrobromide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.