missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzoic Acid, TRC

Product Code. 30135565
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135565 50 g 50g
30136701 100 g 100g
30008037 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135565 Supplier Toronto Research Chemicals Supplier No. TRCD45168050G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-50-3
Molecular Formula C7 H6 O4
Molecular Weight (g/mol) 154.12
Synonym Protocatechuic acid (8CI), 3,4-Dihydroxybenzoic acid, 1,2-Dihydroxybenzene-4-carboxylic acid, 4,5-Dihydroxybenzoic acid, 4-Carboxy-1,2-dihydroxybenzene, NSC 16631, Protocatechoic acid
IUPAC Name 3,4-dihydroxybenzoic acid
SMILES OC(=O)c1ccc(O)c(O)c1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H6O4/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,8-9H,(H,10,11)
Formula Weight 154.0266
Chemical Name or Material Protocatechuic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.