missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzoic acid, 97%

Product Code. 13438308 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13438308 500 g Glass bottle 500g
10714991 25 g Glass bottle 25g
10162810 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13438308 Supplier Thermo Scientific Acros Supplier No. 114895000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-50-3 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-50-3
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002509
InChI Key YQUVCSBJEUQKSH-UHFFFAOYSA-N
Synonym protocatechuic acid, 4-carboxy-1,2-dihydroxybenzene, protocatehuic acid, benzoic acid, 3,4-dihydroxy, protocatechuate, 4,5-dihydroxybenzoic acid, 3,4-dihydroxybenzoicacid, unii-36r5qj8l4b, ccris 6291, chembl37537
PubChem CID 72
ChEBI CHEBI:36062
IUPAC Name 3,4-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (HO)2C6H3CO2H
Quantity 500 g
Beilstein 10,389
Merck Index 15,8003
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol and ether
Formula Weight 154.12
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3, 4-Dihydroxybenzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dihydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.