missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 11424603
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
139-85-5
C7H6O3
138.122
MFCD00003370
IBGBGRVKPALMCQ-UHFFFAOYSA-N
protocatechualdehyde, rancinamycin iv, protocatechuic aldehyde, benzaldehyde, 3,4-dihydroxy, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-dihydroxybenzene, 3,4-dihydroxy-benzaldehyde, 4-formyl-1,2-benzenediol, 1,2-dihydroxy-4-formylbenzene, unii-4pvp2hch4t
8768
CHEBI:50205
3,4-dihydroxybenzaldehyde
C1=CC(=C(C=C1C=O)O)O
This item is not returnable. View return policy
139-85-5
C7H6O3
138.122
MFCD00003370
IBGBGRVKPALMCQ-UHFFFAOYSA-N
protocatechualdehyde, rancinamycin iv, protocatechuic aldehyde, benzaldehyde, 3,4-dihydroxy, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-dihydroxybenzene, 3,4-dihydroxy-benzaldehyde, 4-formyl-1,2-benzenediol, 1,2-dihydroxy-4-formylbenzene, unii-4pvp2hch4t
8768
CHEBI:50205
3,4-dihydroxybenzaldehyde
C1=CC(=C(C=C1C=O)O)O

3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.

Solubility
Soluble in water(6.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 152°C to 157°C
Quantity 5 g
Beilstein 774381
Sensitivity Air Sensitive
Merck Index 14,7893
Solubility Information Soluble in water(6.3g/L).
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material 3,4-Dihydroxybenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'