missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzaldehyde, 98%

Product Code. 11434603
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434603 25 g 25g
11424603 5 g 5g
11444603 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434603 Supplier Thermo Scientific Alfa Aesar Supplier No. A11558.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 139-85-5 | C7H6O3 | 138.122 g/mol

3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.

Solubility
Soluble in water(6.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-85-5
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.122
MDL Number MFCD00003370
InChI Key IBGBGRVKPALMCQ-UHFFFAOYSA-N
Synonym protocatechualdehyde, rancinamycin iv, protocatechuic aldehyde, benzaldehyde, 3,4-dihydroxy, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-dihydroxybenzene, 3,4-dihydroxy-benzaldehyde, 4-formyl-1,2-benzenediol, 1,2-dihydroxy-4-formylbenzene, unii-4pvp2hch4t
PubChem CID 8768
ChEBI CHEBI:50205
IUPAC Name 3,4-dihydroxybenzaldehyde
SMILES C1=CC(=C(C=C1C=O)O)O

Specifications

Melting Point 152°C to 157°C
Quantity 25 g
Beilstein 774381
Sensitivity Air Sensitive
Merck Index 14,7893
Solubility Information Soluble in water(6.3g/L).
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material 3,4-Dihydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.