missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3,4-Dihydroxy-DL-phenylalanine, 98%

Product Code. 11354288
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354288 25 g 25g
11364288 5 g 5g
11344288 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354288 Supplier Thermo Scientific Chemicals Supplier No. 041535.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63-84-3 | C9H11NO4 | 197.19 g/mol

3,4-Dihydroxy-DL-phenylalanine is used as a dopamine precursor. It is also useful in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-84-3
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00063060
InChI Key WTDRDQBEARUVNC-UHFFFAOYNA-N
Synonym dl-dopa, 3,4-dihydroxy-dl-phenylalanine, 2-amino-3-3,4-dihydroxyphenyl propanoic acid, dl-dioxyphenylalanine, dl-dihydroxyphenylalanine, 3-3,4-dihydroxyphenyl-dl-alanine, 3-hydroxytyrosine, 3-hydroxy-dl-tyrosine, dopa, dl-3,4-dopa
PubChem CID 836
ChEBI CHEBI:49168
IUPAC Name 2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

Melting Point 290°C (decomposition)
Merck Index 14,3420
Solubility Information Soluble in water,0.5M dilute hydrochloric acid and formic acid. Insoluble in ethanol,benzene,chloroform and ethyl acetate.
Quantity 25 g
Odor Odorless
Formula Weight 197.19
Sensitivity Air Sensitive
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3,4-Dihydroxy-DL-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.