missing translation for 'onlineSavingsMsg'
Learn More

(-)-3,4-Dihydroxy-alpha-methyl-L-phenylalanine sesquihydrate, 99%

Product Code. 15422846
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422846 1 g 1g
15432846 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422846 Supplier Thermo Scientific Alfa Aesar Supplier No. H56704.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41372-08-1 | C10H15NO5 | 229.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41372-08-1
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.232
MDL Number MFCD00149280
InChI Key XDJUBZFLFDODNF-PPHPATTJSA-N
Synonym methyldopa hydrate, l-methyldopa, s-2-amino-3-3,4-dihydroxyphenyl-2-methylpropionic acid sesquihydrate, --3-3,4-dihydroxyphenyl-2-methyl-l-alanine sesquihydrate, l---alpha-methyldopa hydrate, alpha-methyl-l-dopa sesquihydrate, --3,4-dihydroxy-^a-methyl-l-phenylalanine sesquihydrate, --3-3,4-dihydroxyphenyl-2-methyl-l-alanine ses, 2s-2-amino-3-3,4-dihydroxyphenyl-2-methylpropanoic acid, oxamethane
PubChem CID 18530292
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid;hydrate
SMILES CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)N.O

Specifications

Melting Point >300°C
Merck Index 14,6055
Quantity 1 g
Formula Weight 238.24 (211.22 Anhydrous)
Optical Rotation −13° (c=1 in Water)
Sensitivity Air and light sensitive
Percent Purity 99%
Chemical Name or Material (-)-3,4-Dihydroxy-alpha-methyl-L-phenylalanine sesquihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.