missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxy-3-cyclobutene-1,2-dione, 99%

Product Code. 10209553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209553 100 g Glass bottle 100g
10675012 5 g Glass bottle 5g
10756391 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10209553 Supplier Thermo Scientific Acros Supplier No. 154021000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2892-51-5 | C4H2O4 | 114.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2892-51-5
MDL Number MFCD00001334
InChI Key PWEBUXCTKOWPCW-UHFFFAOYSA-N
Synonym squaric acid, 3,4-dihydroxy-3-cyclobutene-1,2-dione, quadratic acid, 3-cyclobutene-1,2-dione, 3,4-dihydroxy, quadratsaeure, cyclobutenedione, dihydroxy, 1,2-diketo-3,4-dihydroxycyclobutene, dihydroxycyclobutenedione, unii-svr9d0vodw, dihydroxycyclobut-3-ene-1,2-dione
PubChem CID 17913
ChEBI CHEBI:52141
IUPAC Name 3,4-dihydroxycyclobut-3-ene-1,2-dione
SMILES C1(=C(C(=O)C1=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White to Beige
Density 190g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C4H2O4
Quantity 100 g
Beilstein 08, IV, 2701
Fieser 04,466; 13,284; 15,296
Merck Index 15, 8897
Molecular Weight (g/mol) 114.056
Formula Weight 114.06
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dihydroxy-3-cyclobutene-1, 2-dione
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dihydroxy-3-cyclobutene-1,2-dione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.