missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro Naratriptan Oxalate, TRC

Product Code. 30380429
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380429 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380429 Supplier Toronto Research Chemicals Supplier No. TRCD44920225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dihydro Naratriptan Oxalate
CAS 121679-21-8
Molecular Formula C17H23N3O2S . C2H2O4
Quantity 25 mg
Synonym (Naratriptan Impurity B), N-Methyl-3-(1,2,3,6-tetrahydro-1-methyl-4-pyridinyl)-1H-indole-5-ethanesulfonamide Ethanedioate
InChI Formula InChI=1S/C17H23N3O2S.C2H2O4/c1-18-23(21,22)10-7-13-3-4-17-15(11-13)16(12-19-17)14-5-8-20(2)9-6-14;3-1(4)2(5)6/h3-5,11-12,18-19H,6-10H2,1-2H3;(H,3,4)(H,5,6)
SMILES CNS(=O)(=O)CCc1ccc2[nH]cc(C3=CCN(C)CC3)c2c1.OC(=O)C(=O)O
Molecular Weight (g/mol) 423.5
Formula Weight 423.1464
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.