missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-6,7-dimethoxy-4-methyl-3-oxo-2-quinoxalinepropanoic Acid Methyl Ester, TRC

Product Code. 30371483
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371483 50 mg 50mg
30387406 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30371483 Supplier Toronto Research Chemicals Supplier No. TRCD44907550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dihydro-6,7-dimethoxy-4-methyl-3-oxo-2-quinoxalinepropanoic Acid Methyl Ester
CAS 131426-28-3
Molecular Formula C15H18N2O5
Quantity 50 mg
Synonym 6,7-Dimethoxy-4-methyl-3-oxo-3,4-dihydroquinoxaline-2-propionic Acid Methyl Ester
InChI Formula InChI=1S/C15H18N2O5/c1-17-11-8-13(21-3)12(20-2)7-10(11)16-9(15(17)19)5-6-14(18)22-4/h7-8H,5-6H2,1-4H3
SMILES COC(=O)CCC1=Nc2cc(OC)c(OC)cc2N(C)C1=O
IUPAC Name methyl 3-(6,7-dimethoxy-4-methyl-3-oxoquinoxalin-2-yl)propanoate
Molecular Weight (g/mol) 306.31
Formula Weight 306.1216
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.