missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-3-oxo-2-pyrazinecarboxamide, TRC

Product Code. 30371930
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30371930 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371930 Supplier Toronto Research Chemicals Supplier No. TRCD4541501G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 55321-99-8
Molecular Formula C5H5N3O2
Quantity 1 g
Synonym 2-Pyrazinecarboxamide, 3,4-dihydro-3-oxo-, Pyrazinamide, 3-hydroxy- (6CI), Pyrazinecarboxamide, 3,4-dihydro-3-oxo- (9CI), 3,4-Dihydro-3-oxo-2-pyrazinecarboxamide, 3-Hydroxy-2-pyrazinecarboxamide, NSC 163503, T 1105, 3,4-Dihydro-3-oxopyrazine-2-carboxamide, 3-Hydroxypyrazinamide, 3,4-Dihydro-3-oxo-pyrazinecarboxamide, 3-Hydroxy-2-pyrazinecarboxamide, NSC 163503, T 1105
InChI Formula InChI=1S/C5H5N3O2/c6-4(9)3-5(10)8-2-1-7-3/h1-2H,(H2,6,9)(H,8,10)
SMILES NC(=O)C1=NC=CNC1=O
IUPAC Name 2-oxo-1H-pyrazine-3-carboxamide
Molecular Weight (g/mol) 139.11
Formula Weight 139.0382
Chemical Name or Material 3,4-Dihydro-3-oxo-2-pyrazinecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.