missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran, 99%

Artikelnummer. 11327815
Ansicht ändern
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packungsgröße:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
11327815 500 mL 500mL
11307815 25 mL 25mL
11317815 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11327815 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. L02731.AP
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 110-87-2 | C5H8O | 84.118 g/mol

Widely used hydroxyl-protecting reagent3,4-Dihydro-2H-pyran is used as a hydroxyl-protecting reagent in organic synthesis. It acts as an intermediate in synthetic chemistry. It is used to protect various reactive functional groups. It is involved in the polymerization reaction either alone or with unsaturated compound and finds application in polymer industries. Further, it is employed in the preparation of bicyclic compounds of epoxide-fused, halo compounds and allenic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Widely used hydroxyl-protecting reagent3,4-Dihydro-2H-pyran is used as a hydroxyl-protecting reagent in organic synthesis. It acts as an intermediate in synthetic chemistry. It is used to protect various reactive functional groups. It is involved in the polymerization reaction either alone or with unsaturated compound and finds application in polymer industries. Further, it is employed in the preparation of bicyclic compounds of epoxide-fused, halo compounds and allenic alcohols.

Solubility
Miscible with water and chloroform.

Notes
Incompatible with oxidizing agents, strong acids and alcohols.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 110-87-2
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006558
InChI Key BUDQDWGNQVEFAC-UHFFFAOYSA-N
Synonym dihydropyran, 3,4-dihydropyran, 2,3-dihydropyran, 2h-3,4-dihydropyran, 2,3-dihydro-4h-pyran, dihydro-2h-pyran, 2h-pyran, dihydro, 2h-pyran, 3,4-dihydro, 5,6-dihydro-4h-pyran, dihydropyrane
PubChem CID 8080
IUPAC Name 3,4-dihydro-2H-pyran
SMILES C1CC=COC1

Spezifikation

Melting Point -70°C
Density 0.926
pH 7.0
Boiling Point 85°C to 86°C
Flash Point −15°C (5°F)
Odor Unpleasant
Refractive Index 1.442
Quantity 500 mL
UN Number UN2376
Beilstein 103493
Solubility Information Miscible with water and chloroform.
Formula Weight 84.12
Percent Purity 99%
Chemical Name or Material 3,4-Dihydro-2H-pyran
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.