missing translation for 'onlineSavingsMsg'
Learn More

(3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol, 97%, Thermo Scientific™

Product Code. 15447745
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447745 250 mg 250mg
11422070 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447745 Supplier Thermo Scientific Alfa Aesar Supplier No. H28506.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304669-35-0
Molecular Formula C7H14O2Si
Molecular Weight (g/mol) 158.272
MDL Number MFCD09265116
InChI Key ZMDNGTLKIVNNCF-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-pyran-6-yl dimethylsilanol, 5,6-dihydro-4h-pyran-2-yl dimethylsilanol, 2-5,6-dihydro-4h-pyranyl dimethylsilanol, silanol, 3,4-dihydro-2h-pyran-6-yl dimethyl, 5,6-dihydro-4h-pyran-2-yldimethylsilanol, amtsi069
PubChem CID 11332616
IUPAC Name 3,4-dihydro-2H-pyran-6-yl-hydroxy-dimethylsilane
SMILES C[Si](C)(C1=CCCCO1)O

Specifications

Boiling Point 54°C (0.8mmHg)
Quantity 250 mg
Flash Point 91°C (196°F)
Formula Weight 158.27
Percent Purity 97%
Refractive Index 1.475
Density 1.005
Chemical Name or Material (3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.