missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxylic acid, 98%

Product Code. 11416330
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416330 5 g 5g
11406330 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416330 Supplier Thermo Scientific Alfa Aesar Supplier No. H27175.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33632-74-5 | C10H10O4 | 194.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33632-74-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00276310
InChI Key BYYFEADAQZCIGM-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepine-2-carboxylic acid, 2h-1,5-benzodioxepin-2-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepine-2-carboxylic acid, 2h-1,5-benzodioxepin-2-carboxylic acid, 3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylicacid
PubChem CID 53708627
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-4-carboxylic acid
SMILES C1COC2=CC=CC=C2OC1C(=O)O

Specifications

Melting Point 146°C to 148°C
Quantity 5 g
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.